Learn More
BioMed Diagnostics Nylon tip, Individually sealed, sterile, flocked swabs with breakable shaft. For collecting SARS- CoV-2 and other specimens. | Detects: collection of Nasopharyngeal specimens. | Available in: 100 pack
Catalog No. NC2582823
Encompass
Nylon tip, Individually sealed, sterile, flocked swabs with plastic, breakable shaft. For collecting SARS- CoV-2 and other clinical specimens. | Product Principle: 1.) Proprietary nylon tip flocking process optimizes specimens collection and instant, spontaneous elution in media with up to 95% release. 2.) No-slip, frosted, flexible plastic handle with molded breakpoint, ergonomic & anatomic designs for optimal patient comfort and efficient sample collection. 3.) Compatible with most diagnostic molecular and antigen tests. | Simple Convenience: 80 mm breakpoint (fits most standard 5mL, 10mL, 12mL, 15mL tubes) | Detection: For the collection of Nasopharyngeal specimens, including SARS-CoV-2. | Available in: 100 pack
This item is not returnable. View return policy
Catalog No. NC2582823 Supplier BioMed Diagnostics Supplier No. 10000040
Nylon tip, Individually sealed, sterile, flocked swabs with plastic, breakable shaft. For collecting SARS- CoV-2 and other clinical specimens. | Product Principle: 1.) Proprietary nylon tip flocking process optimizes specimens collection and instant, spontaneous elution in media with up to 95% release. 2.) No-slip, frosted, flexible plastic handle with molded breakpoint, ergonomic & anatomic designs for optimal patient comfort and efficient sample collection. 3.) Compatible with most diagnostic molecular and antigen tests. | Simple Convenience: 80 mm breakpoint (fits most standard 5mL, 10mL, 12mL, 15mL tubes) | Detection: For the collection of Nasopharyngeal specimens, including SARS-CoV-2. | Available in: 100 pack