Learn More
Medchemexpress LLC Iclepertin | 1421936-85-7 | MFCD34474019 | 100.0% | C20H18F6N2O5S | 10MG
SDP
Catalog No. 5000260318
Encompass_Preferred
Iclepertin is a potent, selective, orally active glycine transporter 1 (GlyT1) inhibitor provided for research use. The compound elevates extracellular glycine and has been investigated for cognitive impairment in neurological disorders; it is intended only for laboratory research and not for human administration.
  • Potent, selective glycine transporter 1 (GlyT1) inhibitor.
  • Orally active; relevant for in vivo and in vitro studies.
  • High chemical purity (99.97%).
  • Used in preclinical research on cognitive impairment and schizophrenia.
  • Available in small lab-scale pack sizes suitable for experimental workflows.
Catalog No. 50-002-60318 Supplier Medchemexpress LLC Supplier No. HY13893510MG
Iclepertin is a potent, selective, orally active glycine transporter 1 (GlyT1) inhibitor provided for research use. The compound elevates extracellular glycine and has been investigated for cognitive impairment in neurological disorders; it is intended only for laboratory research and not for human administration.
  • Potent, selective glycine transporter 1 (GlyT1) inhibitor.
  • Orally active; relevant for in vivo and in vitro studies.
  • High chemical purity (99.97%).
  • Used in preclinical research on cognitive impairment and schizophrenia.
  • Available in small lab-scale pack sizes suitable for experimental workflows.