Learn More
Cellular Dynamics International iCell Cardiomyocytes², 01434 CardioTox Kit, 5M cells
Catalog No. NC2949631 Shop All Cellular Dynamics International Products
Encompass
iCell Cardiomyocytes2 from FUJIFILM Cellular Dynamics provides relevant biology and rapid workflows making them powerful tools for safety, toxicology, and drug discovery research. FCDI offers a full suite of cardiomyocyte media that are all DMEM-based with the same physiological ion concentrations and mature glucose-free energy source. Rigorously designed and quality controlled to work with iCell Cardiomyocytes and Cardiomyocytes2, these media provide researchers the ability to seamlessly switch between serum-containing, serum-free, and albumin-free conditions, enabling acute and chronic studies.
Catalog No. NC2949631 Supplier Cellular Dynamics International Supplier No. R1222
iCell Cardiomyocytes2 from FUJIFILM Cellular Dynamics provides relevant biology and rapid workflows making them powerful tools for safety, toxicology, and drug discovery research. FCDI offers a full suite of cardiomyocyte media that are all DMEM-based with the same physiological ion concentrations and mature glucose-free energy source. Rigorously designed and quality controlled to work with iCell Cardiomyocytes and Cardiomyocytes2, these media provide researchers the ability to seamlessly switch between serum-containing, serum-free, and albumin-free conditions, enabling acute and chronic studies.