Learn More
Medchemexpress LLC Icomidocholic acid (Aramchol) | 246529-22-6 | 99.98% | 702.10 | 1 ML
SDP
Catalog No. 5000402062
Encompass_Preferred
Icomidocholic acid (Aramchol) is a lipid molecule synthesized from cholic acid and arachidic acid. It is an orally active SCD1 inhibitor and cholesterol solubilizer with antifibrotic effects. Icomidocholic acid can reduce liver fat content, dissolve cholesterol crystals, and prevent gallstone formation, making it suitable for the study of nonalcoholic fatty liver disease (NAFLD) and nonalcoholic steatohepatitis (NASH).
  • Orally active SCD1 inhibitor
  • Cholesterol solubilizer
  • Antifibrotic effects
  • Reduces liver fat content
  • Dissolves cholesterol crystals
  • Prevents gallstone formation
  • Suitable for NAFLD and NASH studies
Catalog No. 50-004-02062 Supplier Medchemexpress LLC Supplier No. HY1979610MM/1ML
Add to Cart
Add to Cart
Icomidocholic acid (Aramchol) is a lipid molecule synthesized from cholic acid and arachidic acid. It is an orally active SCD1 inhibitor and cholesterol solubilizer with antifibrotic effects. Icomidocholic acid can reduce liver fat content, dissolve cholesterol crystals, and prevent gallstone formation, making it suitable for the study of nonalcoholic fatty liver disease (NAFLD) and nonalcoholic steatohepatitis (NASH).
  • Orally active SCD1 inhibitor
  • Cholesterol solubilizer
  • Antifibrotic effects
  • Reduces liver fat content
  • Dissolves cholesterol crystals
  • Prevents gallstone formation
  • Suitable for NAFLD and NASH studies