Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Tingley Rain Pants, Polyester, Snaps Closure, Black, X-Large, 1 ea

Catalog No. 5000673211
Encompass

These rain pants are designed to provide waterproof protection. Constructed from durable polyester, they feature a snap closure and are suitable for various outdoor conditions.

  • Pants style
  • X-large size
  • Black color
  • Inseam length: 31 in.
  • Non-hazardous protection
  • Hook & loop ankle
  • Waist size: 44 to 46 in.
  • Material weight: 5.5 oz.
  • Snap fly
  • Top-stitched and taped seams
  • Material thickness: 0.30 mm

Catalog No. 50-006-73211 Supplier MSC Supplier No. 31659915
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These rain pants are designed to provide waterproof protection. Constructed from durable polyester, they feature a snap closure and are suitable for various outdoor conditions.

  • Pants style
  • X-large size
  • Black color
  • Inseam length: 31 in.
  • Non-hazardous protection
  • Hook & loop ankle
  • Waist size: 44 to 46 in.
  • Material weight: 5.5 oz.
  • Snap fly
  • Top-stitched and taped seams
  • Material thickness: 0.30 mm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.