Learn More
MSC Tingley Icon Jacket, Polyurethane on 300 Denier Polyester, Small, 1 ea
Catalog No. 5000685423
Encompass
This general-purpose jacket is constructed from 300 denier polyester with a breathable polyurethane coating, providing 100% waterproof protection. Designed for utilities and construction work, the garment features a dark color to help conceal dirt and an overhead shoulder design to ensure freedom of movement.
  • Stitched and taped sealed seams for waterproof performance
  • Zipper and storm fly front closure with an integrated throat guard
  • Roll-a-way adjustable hood integrated into the collar
  • Equipped with 7 pockets, including cargo, hand warmer, chest, and pencil pockets
  • Elastic cuffs with hook and loop straps for a secure fit
  • Adjustable elastic drawcord at the sweep
  • Compatible as a shell with system-specific liners
  • Tested via ASTM E96/E96M for water vapor transmission
Catalog No. 50-006-85423 Supplier MSC Supplier No. 40085599
This general-purpose jacket is constructed from 300 denier polyester with a breathable polyurethane coating, providing 100% waterproof protection. Designed for utilities and construction work, the garment features a dark color to help conceal dirt and an overhead shoulder design to ensure freedom of movement.
  • Stitched and taped sealed seams for waterproof performance
  • Zipper and storm fly front closure with an integrated throat guard
  • Roll-a-way adjustable hood integrated into the collar
  • Equipped with 7 pockets, including cargo, hand warmer, chest, and pencil pockets
  • Elastic cuffs with hook and loop straps for a secure fit
  • Adjustable elastic drawcord at the sweep
  • Compatible as a shell with system-specific liners
  • Tested via ASTM E96/E96M for water vapor transmission