Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human ICOS (CD278) ELISA Kit

Catalog No. EHICOSX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EHICOSX5 5 x 96 Tests
EHICOS 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EHICOSX5 Supplier Invitrogen™ Supplier No. EHICOSX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

The Human ICOS (CD278) ELISA quantitates Hu ICOS in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu ICOS. Principle of the method The Human ICOS solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

ICOS (CD278) encodes a protein that is a member of the CD28 and CTLA-4 cell-surface receptor family. It functions as a homodimer and has a crucial role in cell-cell signaling, immune responses, and the regulation of cell proliferation. ICOS has been linked to various disorders, including Immunodeficiency, Common Variable, 1 and Common Variable Immunodeficiency.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9Y6W8
Assay Range 1.229-300 ng/mL
Assay Sensitivity 1.2 ng/mL
Calibration Range 1.2 ng/mL to 300 ng/mL
Assay Solid-phase sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Cell Structure/Organelle Proteins
Sample Type Plasma, Serum, Supernatant
Specificity This ELISA pair antibody detects human ICOS. Other species not determined.
Sufficient For 5 x 96 Assays
Target Species Human
For Use With (Application) ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias Activation-inducible lymphocyte immunomediatory molecule; Ailim; CCLP; CD278; CD28 and CTLA-4-like protein; CD28-related protein 1; CRP-1; CVID1; H4; Icos; inducible costimulator; inducible T cell costimulator; inducible T cell co-stimulator; inducible T-cell costimulator; inducible T-cell co-stimulator; Ly115Activation-inducible lymphocyte immunomediatory molecule; Ailim; CCLP; CD278; CD28 and CTLA-4-like protein; CD28-related protein 1; CRP-1; CVID1; H4; Icos; inducible costimulator; inducible T cell costimulator; inducible T cell co-stimulator; inducible T-cell costimulator; inducible T-cell co-stimulator; Ly115; AILIM; CD278; CVID1
Gene ID (Entrez) 29851
Gene Symbol ICOS
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Protein Aliases Activation-inducible lymphocyte immunomediatory molecule
Quantity 5 x 96 Tests
Regulatory Status RUO
Research Discipline Extracellular Matrix & Cell Adhesion
Sample Volume Plasma 25 μL, Serum 25 μL, Supernatant 100 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Synonym AILIM, CD278, CVID1
Target ICOS (CD278)
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.