Learn More
ACROBiosystems ACROBiosystems Human ICOS / CD278 Protein, Fc Tag
Catalog No. 501957014
Encompass_Preferred
Recombinant Protein;1MG;Human ICOS, Fc Tag (ICS-H5258) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Phe 141 (Accession # Q9Y6W8-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 40.2 kDa. The protein migrates as 46-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human B7-H2, His Tag (Cat. No. B72-H5221) at 1 μg/mL (100 μL/well) can bind Human ICOS, Fc Tag (Cat. No. ICS-H5258) with a linear range of 0.4-6 ng/mL (QC tested).
Catalog No. 50-195-7014 Supplier ACROBiosystems Supplier No. ICSH52581MG
Recombinant Protein;1MG;Human ICOS, Fc Tag (ICS-H5258) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Phe 141 (Accession # Q9Y6W8-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 40.2 kDa. The protein migrates as 46-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human B7-H2, His Tag (Cat. No. B72-H5221) at 1 μg/mL (100 μL/well) can bind Human ICOS, Fc Tag (Cat. No. ICS-H5258) with a linear range of 0.4-6 ng/mL (QC tested).