Learn More
Genscript Corporation ICOS Fc Chimera Human
Supplier:  Genscript Corporation Z03412100
Encompass
Inducible T-cell costimulator is an immune checkpoint protein that in humans is encoded by the ICOS gene CD278 or ICOS (Inducible T-cell COStimulator) is a CD28-superfamily costimulatory molecule that is expressed on activated T cells It is thought to be important for Th2 cells in particular The protein encoded by this gene belongs to the CD28 and CTLA-4 cell-surface receptor family It forms homodimers and plays an important role in cell-cell signaling immune responses and regulation of cell proliferation
Catalog No. 50-000-59521