Learn More
ACROBiosystems ACROBiosystems Mouse ICOS / CD278 (C137S, C138S) Protein, Fc Tag
Supplier: ACROBiosystems ICSM5259100UG
Recombinant Protein;100UG;Mouse ICOS (C137S, C138S), Fc Tag (ICS-M5259) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Leu 144 (Accession # Q9WVS0-1 (C137S, C138S)).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 40.6 kDa. The protein migrates as 45-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human B7-H2, His Tag (Cat. No. B72-H5221) at 1 μg/mL (100 μL/well) can bind Mouse ICOS (C137S, C138S), Fc Tag (Cat. No. ICS-M5259) with a linear range of 0.3-10 ng/mL (QC tested).