Learn More
ACROBiosystems ACROBiosystems Biotinylated Human ICOS / CD278 (C136S, C137S) Protein, His,Avitag™ (recommended for biopanning)
Supplier: ACROBiosystems ICSH82E525UG
Recombinant Protein;25UG;Biotinylated Human ICOS (C136S, C137S), His,Avitag (recommended for biopanning) (ICS-H82E5) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Phe 141 (Accession # Q9Y6W8-1 (C136S, C137S)).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 17.4 kDa. The protein migrates as 25-32 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human B7-H2, Fc Tag (Cat. No. B72-H5254) at 5 μg/mL (100 μL/well) can bind Biotinylated Human ICOS (C136S, C137S), His,Avitag (recommended for biopanning) (Cat. No. ICS-H82E5) with a linear range of 2-78 ng/mL (QC tested).