Learn More
ACROBiosystems ACROBiosystems Mouse ICOS / CD278 (C137S, C138S) Protein, His Tag
Supplier:  ACROBiosystems ICSM52H81MG
Encompass_Preferred
Recombinant Protein;1MG;Mouse ICOS (C137S, C138S), His Tag (ICS-M52H8) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Leu 144 (Accession # Q9WVS0-1 (C137S,C138S)).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 15.7 kDa. The protein migrates as 22-30 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human B7-H2, Fc Tag (Cat. No. B72-H5254) at 1 μg/mL (100 μL/well) can bind Mouse ICOS (C137S, C138S), His Tag (Cat. No. ICS-M52H8) with a linear range of 0.01-0.2ng/mL (QC tested).
Catalog No. 50-195-6918