Learn More
Sino Biological Human ICOS Ligand / B7-H2 / ICOSLG Protein (His Tag), Biotinylated 20ug
Supplier:  Sino Biological 11559H08HB20
Encompass_Preferred
A DNA sequence encoding the human ICOSLG (NP_056074.1) (Met1-Ser258) was expressed with a C-terminal polyhistidine tag. The purified protein was biotinylated in vitro.
Catalog No. 50-211-6029