Learn More
Medchemexpress LLC ICOS Protein Mouse 10ug | 10ug
SDP
Supplier:  Medchemexpress LLC HYP7867310UG
Encompass_Preferred
ICOS proteins optimize T cell responses to foreign antigens promoting proliferation lymphokine secretion upregulation of cell-cell interaction molecules and efficient B cell antibody secretion ICOS is essential for efficient communication of T cells and B cells and for normal antibody responses to T cell-dependent antigens ICOS Protein Mouse (C137S C138S HEK293 Fc) is the recombinant mouse-derived ICOS protein expressed by HEK293 with C-hFc labeled tag and C137S C138S mutation
Catalog No. 50-001-94239