Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Icos protein, rat (HEK293, Fc) | >95.0% | 5 UG
SDP

Supplier:  Medchemexpress LLC HYP748685UG

Encompass_Preferred

Recombinant rat ICOS (inducible T-cell co-stimulator) protein expressed in HEK293 cells and fused to a C-terminal human Fc tag. Supplied lyophilized from PBS (pH 7.4), the glycosylated Fc-fusion spans residues E21-L142 with an estimated molecular weight of ~43-50 kDa. Designed for research into T cell co-stimulation and immune signaling.

  • Purity >95% by reducing SDS-PAGE.
  • Contains C-terminal human Fc tag for detection and capture.
  • Supplied lyophilized for extended shelf life and transport.
  • Expressed in HEK293 cells to provide mammalian glycosylation.
  • Amino acid range E21-L142 corresponds to extracellular domain.
  • Recommended storage at -20°C; reconstituted material stable at 4°C for 1 week.
  • Available in research-scale sizes including a 5 μg package.

Catalog No. 50-002-93728


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.