Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC ICOS protein, rat (HEK293, Fc) | >95.0% | 50 UG
SDP

Catalog No. 5000291225
Encompass_Preferred

Recombinant rat ICOS extracellular domain fused to a C-terminal human Fc, produced in HEK293 cells and supplied lyophilized. It is intended for immunology and cell-signaling research, including receptor-ligand binding assays and antibody characterization.

  • Recombinant rat ICOS extracellular domain with C-terminal human Fc tag.
  • Expressed in HEK293 mammalian cells for native glycosylation.
  • Sequence corresponds to residues E21-L142 (accession Q9R1T7-1).
  • High purity >95.0% by reducing SDS-PAGE.
  • Lyophilized from 0.22 μm filtered PBS, pH 7.4; store at -20°C.
  • Available in small research quantities (for example, 50 μg) for assay use.

Catalog No. 50-002-91225 Supplier Medchemexpress LLC Supplier No. HYP7486850UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant rat ICOS extracellular domain fused to a C-terminal human Fc, produced in HEK293 cells and supplied lyophilized. It is intended for immunology and cell-signaling research, including receptor-ligand binding assays and antibody characterization.

  • Recombinant rat ICOS extracellular domain with C-terminal human Fc tag.
  • Expressed in HEK293 mammalian cells for native glycosylation.
  • Sequence corresponds to residues E21-L142 (accession Q9R1T7-1).
  • High purity >95.0% by reducing SDS-PAGE.
  • Lyophilized from 0.22 μm filtered PBS, pH 7.4; store at -20°C.
  • Available in small research quantities (for example, 50 μg) for assay use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.