Learn More
Cayman Chemical ICOSCD278CTerm RabbIt Mo 100uL
SDP
Catalog No. 502948547
Encompass_Preferred
A mutagenic heterocyclic amine mutagenic to S. typhimurium in the Ames test induces tumor formation in the Zymbal gland oral cavity and colon in male and female rats the skin of male rats and the mammary gland in female rats at 0.03 in the diet
Catalog No. 50-294-8547 Supplier Cayman Chemical Supplier No. 32338100 UL
A mutagenic heterocyclic amine mutagenic to S. typhimurium in the Ames test induces tumor formation in the Zymbal gland oral cavity and colon in male and female rats the skin of male rats and the mammary gland in female rats at 0.03 in the diet