Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ProSci ICOSLG Peptide
SDP

Catalog No. 16595480
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-595-480 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-595-480 Supplier ProSci Supplier No. 8687P0050
Only null left
Add to Cart
Add to Cart

Used for Blocking Activity

ICOSLG peptide is used for blocking the activity of ICOSLG antibody.

Specifications

Gene ID (Entrez) 23308
Species Human
Accession Number O75144
Common Name inducible T cell costimulator ligand
Concentration 200 μg/mL
Content And Storage Store ICOSLG peptide at -20°C, stable for one year.
Format Liquid
Formulation PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide
For Use With (Application) Blocking Assay
Gene Symbol ICOSLG
Quantity 0.05 mg
Sequence 17 amino acids near the center of human ICOSLG.
Synonym GL50, Inducible T Cell Costimulator Ligand, B7-Related Protein, B7 Homolog, Inducible T-Cell Costimulator Ligand, B7-Like Protein Gl50, B7 Homologue, B7RP-1, ICOSL, B7-H2, B7RP1, B7H2, Transmembrane Protein B7-H2 ICOS Ligand, Inducible T-Cell Co-Stimulator Ligand, CD275 Antigen, ICOS Ligand, KIAA0653, ICOS-L, CD275, LICOS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.