Learn More
Selleck Chemical LLC Icotinib (BPI-2009H) S2922-200mg
SDP
Catalog No. 503109602
Encompass_Preferred
Icotinib (BPI-2009H) is a potent and specific EGFR inhibitor with IC50 of 5 nM including the EGFR EGFR(L858R) EGFR(L861Q) EGFR(T790M) and EGFR(T790M L858R)
Catalog No. 50-310-9602 Supplier Selleck Chemical LLC Supplier No. S2922200MG
Icotinib (BPI-2009H) is a potent and specific EGFR inhibitor with IC50 of 5 nM including the EGFR EGFR(L858R) EGFR(L861Q) EGFR(T790M) and EGFR(T790M L858R)