Learn More
Medchemexpress LLC Icotinib hydrochloride | 1204313-51-8 | C22H22ClN3O4 | 10 MM 1 ML
SDP
Catalog No. 5000373221
Encompass_Preferred
Icotinib Hydrochloride is a potent, CNS-penetrant, and specific Epidermal Growth Factor Receptor (EGFR) inhibitor. It is effective against various mutant forms of EGFR and functions as a click chemistry reagent due to its Alkyne group, enabling copper-catalyzed azide-alkyne cycloaddition with Azide groups. This compound inhibits the proliferation of several cancer cell lines and demonstrates dose-dependent antitumor effects in animal models.
  • Potent and specific EGFR inhibitor
  • Effective against mutant EGFR forms
  • Functions as a click chemistry reagent
  • Inhibits kinase activity and tumor cell proliferation
  • Exhibits dose-dependent antitumor effects in vivo
  • Well-tolerated in animal studies
  • Investigated in clinical trials for Non-Small Cell Lung Cancer and other conditions
Catalog No. 50-003-73221 Supplier Medchemexpress LLC Supplier No. HY1516410MM/1ML
Icotinib Hydrochloride is a potent, CNS-penetrant, and specific Epidermal Growth Factor Receptor (EGFR) inhibitor. It is effective against various mutant forms of EGFR and functions as a click chemistry reagent due to its Alkyne group, enabling copper-catalyzed azide-alkyne cycloaddition with Azide groups. This compound inhibits the proliferation of several cancer cell lines and demonstrates dose-dependent antitumor effects in animal models.
  • Potent and specific EGFR inhibitor
  • Effective against mutant EGFR forms
  • Functions as a click chemistry reagent
  • Inhibits kinase activity and tumor cell proliferation
  • Exhibits dose-dependent antitumor effects in vivo
  • Well-tolerated in animal studies
  • Investigated in clinical trials for Non-Small Cell Lung Cancer and other conditions