Learn More
Medchemexpress LLC Icotinib hydrochloride | 1204313-51-8 | C22H22ClN3O4 | 25 MG
SDP
Catalog No. 5000374236
Encompass_Preferred
Icotinib hydrochloride is a potent and specific EGFR inhibitor that is CNS-penetrant. It is effective against wild-type and various mutant EGFR forms, including EGFRL858R, EGFRL858R/T790M, EGFRT790M, and EGFRL861Q. This compound also functions as a click chemistry reagent, featuring an alkyne group that enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules.
  • Potent and specific EGFR inhibitor
  • CNS-penetrant
  • Inhibits mutant EGFR forms: EGFRL858R, EGFRL858R/T790M, EGFRT790M, and EGFRL861Q
  • Click chemistry reagent with an alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
Catalog No. 50-003-74236 Supplier Medchemexpress LLC Supplier No. 25MGHY15164
Icotinib hydrochloride is a potent and specific EGFR inhibitor that is CNS-penetrant. It is effective against wild-type and various mutant EGFR forms, including EGFRL858R, EGFRL858R/T790M, EGFRT790M, and EGFRL861Q. This compound also functions as a click chemistry reagent, featuring an alkyne group that enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules.
  • Potent and specific EGFR inhibitor
  • CNS-penetrant
  • Inhibits mutant EGFR forms: EGFRL858R, EGFRL858R/T790M, EGFRT790M, and EGFRL861Q
  • Click chemistry reagent with an alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)