Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Icotinib hydrochloride | 1204313-51-8 | MFCD22124500 | 99.6% | 427.88 g/mol | C22H22ClN3O4 | 10 MG
SDP

Catalog No. 5000286466
Encompass_Preferred

Icotinib hydrochloride is the hydrochloride salt of icotinib, a quinazoline-based epidermal growth factor receptor (EGFR) tyrosine kinase inhibitor used in preclinical and analytical research. It is supplied as a solid white to off-white powder suitable for in vitro assays and compound characterization.

  • Potent EGFR inhibitor (IC50 ≈ 5 nM).
  • High purity suitable for analytical use (≈99.6%).
  • White to off-white solid powder physical form.
  • Supplied as a 10 mg quantity for small-scale studies.
  • CAS number 1204313-51-8 for unambiguous identification.
  • Storage: store sealed, away from moisture at 4°C; in solvent, -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-86466 Supplier Medchemexpress LLC Supplier No. HY1516410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Icotinib hydrochloride is the hydrochloride salt of icotinib, a quinazoline-based epidermal growth factor receptor (EGFR) tyrosine kinase inhibitor used in preclinical and analytical research. It is supplied as a solid white to off-white powder suitable for in vitro assays and compound characterization.

  • Potent EGFR inhibitor (IC50 ≈ 5 nM).
  • High purity suitable for analytical use (≈99.6%).
  • White to off-white solid powder physical form.
  • Supplied as a 10 mg quantity for small-scale studies.
  • CAS number 1204313-51-8 for unambiguous identification.
  • Storage: store sealed, away from moisture at 4°C; in solvent, -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.