Learn More
Inorganic Ventures 10 000µg/mL BORON 500mL
SDP
Catalog No. 50723207
Encompass_Preferred
Inorganic Ventures' 10,000 ppm (µg/mL) Boron for ICP is a 500mL certified reference material set in a ammonium hydroxide matrix for stability. This boron standard is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. See Certificate of Analysis for expiration.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-207 Supplier Inorganic Ventures Supplier No. CGB10500ML
Inorganic Ventures' 10,000 ppm (µg/mL) Boron for ICP is a 500mL certified reference material set in a ammonium hydroxide matrix for stability. This boron standard is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. See Certificate of Analysis for expiration.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search