Learn More
Inorganic Ventures 10 000µg/mL CHROMIUM 3 125mL
SDP
Catalog No. 50723242
Encompass_Preferred
Matrix: Nitric Acid (HNO3), Starting Material: Cr metal, Analyte: Cr+3 10000 ug/mL.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-242 Supplier Inorganic Ventures Supplier No. CGCR(3)10125ML
Matrix: Nitric Acid (HNO3), Starting Material: Cr metal, Analyte: Cr+3 10000 ug/mL.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search