Learn More
Inorganic Ventures 10 000µg/mL COPPER 125mL
SDP
Catalog No. 50723252
Encompass_Preferred
Matrix: Nitric Acid (HNO3), Starting Material: Cu metal, Analyte: Cu 10000 ug/mL.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-252 Supplier Inorganic Ventures Supplier No. CGCU10125ML
Matrix: Nitric Acid (HNO3), Starting Material: Cu metal, Analyte: Cu 10000 ug/mL.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search