Learn More
Inorganic Ventures 10 000µg/mL MOLYBDENUM 125mL
SDP
Catalog No. 50723332
Encompass_Preferred
Inorganic Ventures' 10,000 ppm (µg/mL) Molybdenum for ICP is a 125mL certified reference material set in an ammonium hydroxide matrix for stability. This molybdenum standard is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. See Certificate of Analysis for expiration.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-332 Supplier Inorganic Ventures Supplier No. CGMO10125ML
Inorganic Ventures' 10,000 ppm (µg/mL) Molybdenum for ICP is a 125mL certified reference material set in an ammonium hydroxide matrix for stability. This molybdenum standard is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. See Certificate of Analysis for expiration.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search