Learn More
Inorganic Ventures 10 000µg/mL PHOSPHORUS 125mL
SDP
Catalog No. 50723358
Encompass_Preferred
Matrix: Water (H2O), Starting Material: H3PO4, Analyte: P 10000 ug/mL.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-358 Supplier Inorganic Ventures Supplier No. CGP10125ML
Matrix: Water (H2O), Starting Material: H3PO4, Analyte: P 10000 ug/mL.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search