Learn More
Inorganic Ventures 10 000µg/mL SODIUM 500mL
SDP
Catalog No. 50723341
Encompass_Preferred
Matrix: Nitric Acid (HNO3), Starting Material: Na2CO3, Analyte: Na 10000 ug/mL.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-341 Supplier Inorganic Ventures Supplier No. CGNA10500ML
Matrix: Nitric Acid (HNO3), Starting Material: Na2CO3, Analyte: Na 10000 ug/mL.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search