Learn More
Inorganic Ventures 10 000µg/mL TITANIUM 500mL
SDP
Supplier:  Inorganic Ventures CGTI10500ML
Encompass_Preferred
Matrix: Nitric Acid / Hydrofluoric Acid (HNO3 / HF), Starting Material: Ti metal, Analyte: Ti 10000 ug/mL.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-461