Learn More
Inorganic Ventures 1000µg/mL ARSENIC 3 125mL
SDP
Catalog No. 50723196
Encompass_Preferred
Inorganic Ventures' 1000 ppm (µg/mL) Arsenic+3 for ICP is a 125mL certified reference material set in a hydrochloric (tr. sodium hydroxide & sodium bicarb.) matrix for stability. This arsenic+3 standard is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. See Certificate of Analysis for expiration.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-196 Supplier Inorganic Ventures Supplier No. CGAS(3)1125ML
Inorganic Ventures' 1000 ppm (µg/mL) Arsenic+3 for ICP is a 125mL certified reference material set in a hydrochloric (tr. sodium hydroxide & sodium bicarb.) matrix for stability. This arsenic+3 standard is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. See Certificate of Analysis for expiration.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search