Learn More
Inorganic Ventures 1000µg/mL HAFNIUM 500mL
SDP
Catalog No. 50723287
Encompass_Preferred
Inorganic Ventures' 1000 ppm (µg/mL) Hafnium for ICP is a 500mL certified reference material set in a nitric acid / hydrofluoric acid matrix for stability. This hafnium standard is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. See Certificate of Analysis for expiration.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-287 Supplier Inorganic Ventures Supplier No. CGHF1500ML
Inorganic Ventures' 1000 ppm (µg/mL) Hafnium for ICP is a 500mL certified reference material set in a nitric acid / hydrofluoric acid matrix for stability. This hafnium standard is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. See Certificate of Analysis for expiration.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search