Learn More
Inorganic Ventures 1000µg/mL OSMIUM 125mL
SDP
Catalog No. 50723356
Encompass_Preferred
Matrix: Hydrochloric Acid (HCl), Starting Material: (NH4)2OsCl6, Analyte: Os 1000 ug/mL.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-356 Supplier Inorganic Ventures Supplier No. CGOS1125ML
Matrix: Hydrochloric Acid (HCl), Starting Material: (NH4)2OsCl6, Analyte: Os 1000 ug/mL.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search