Learn More
Inorganic Ventures 1000µg/mL SILICA NaOH 500mL
SDP
Catalog No. 50723429
Encompass_Preferred
Inorganic Ventures' 1000 ppm (µg/mL) Silica for ICP is a 500mL certified reference material set in a sodium hydroxide matrix for stability. This silica standard is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. See Certificate of Analysis for expiration.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-429 Supplier Inorganic Ventures Supplier No. CGSIONA1500ML
Inorganic Ventures' 1000 ppm (µg/mL) Silica for ICP is a 500mL certified reference material set in a sodium hydroxide matrix for stability. This silica standard is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. See Certificate of Analysis for expiration.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search