Learn More
Inorganic Ventures 1000µg/mL TIN 125mL
SDP
Supplier:  Inorganic Ventures CGSN1125ML
Encompass_Preferred
Matrix: Nitric Acid / Hydrofluoric Acid (HNO3 / HF), Starting Material: Sn metal, Analyte: Sn 1000 ug/mL.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-436