Learn More
Inorganic Ventures 1000µg/mL VANADIUM 125mL
SDP
Supplier:  Inorganic Ventures CGV1125ML
Encompass_Preferred
Matrix: Nitric Acid (HNO3), Starting Material: V2O5, Analyte: V 1000 ug/mL.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-476