Learn More
Inorganic Ventures 100µg/mL CADMIUM 125mL
SDP
Catalog No. 50723717
Encompass_Preferred
Matrix: Nitric Acid (HNO3), Starting Material: Cd metal, Analyte: Cd 100 ug/mL.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-717 Supplier Inorganic Ventures Supplier No. MSCD100PPM125ML
Matrix: Nitric Acid (HNO3), Starting Material: Cd metal, Analyte: Cd 100 ug/mL.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search