Learn More
Inorganic Ventures 100µg/mL INDIUM 125mL
SDP
Catalog No. 50723737
Encompass_Preferred
Matrix: Nitric Acid (HNO3), Starting Material: In metal, Analyte: In 100 ug/mL.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-737 Supplier Inorganic Ventures Supplier No. MSIN100PPM125ML
Matrix: Nitric Acid (HNO3), Starting Material: In metal, Analyte: In 100 ug/mL.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search