Learn More
Inorganic Ventures 100µg/mL SULFUR 125mL
SDP
Catalog No. 50723761
Encompass_Preferred
Matrix: Water (H2O), Starting Material: H2SO4, Analyte: S 100 ug/mL.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-761 Supplier Inorganic Ventures Supplier No. MSS100PPM125ML
Matrix: Water (H2O), Starting Material: H2SO4, Analyte: S 100 ug/mL.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search