Learn More
Inorganic Ventures 10µg/mL MERCURY HCl 125mL
SDP
Catalog No. 50723734
Encompass_Preferred
Matrix: Hydrochloric Acid (HCl), Starting Material: Hg metal, Analyte: Hg 10 ug/mL.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-734 Supplier Inorganic Ventures Supplier No. MSHG10PPM125ML
Matrix: Hydrochloric Acid (HCl), Starting Material: Hg metal, Analyte: Hg 10 ug/mL.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search