Learn More
Inorganic Ventures 10µg/mL MOLYBDENUM 125mL
SDP
Catalog No. 50723748
Encompass_Preferred
Matrix: Ammonium Hydroxide (NH4OH), Starting Material: (NH4)2MoO4, Analyte: Mo 10 ug/mL.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-748 Supplier Inorganic Ventures Supplier No. MSMO10PPM125ML
Matrix: Ammonium Hydroxide (NH4OH), Starting Material: (NH4)2MoO4, Analyte: Mo 10 ug/mL.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search