Learn More
Inorganic Ventures 10µg/mL TUNGSTEN 125mL
SDP
Catalog No. 50723789
Encompass_Preferred
Matrix: Nitric Acid / Hydrofluoric Acid (HNO3 / HF), Starting Material: W metal, Analyte: W 10 ug/mL.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-789 Supplier Inorganic Ventures Supplier No. MSW10PPM125ML
Matrix: Nitric Acid / Hydrofluoric Acid (HNO3 / HF), Starting Material: W metal, Analyte: W 10 ug/mL.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search