Learn More
Inorganic Ventures 10µg/mL URANIUM 125mL
SDP
Catalog No. 50723785
Encompass_Preferred
Inorganic Ventures' 10 ppm (µg/mL) Uranium for ICP-MS is a 125mL certified reference material set in a nitric acid matrix for stability. This uranium standard is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. See Certificate of Analysis for expiration.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-785 Supplier Inorganic Ventures Supplier No. MSU10PPM125ML
Inorganic Ventures' 10 ppm (µg/mL) Uranium for ICP-MS is a 125mL certified reference material set in a nitric acid matrix for stability. This uranium standard is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. See Certificate of Analysis for expiration.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search