Learn More
Inorganic Ventures SOIL WATER SPIKE SOLN 2 125mL
SDP
Catalog No. 50723520
Encompass_Preferred
Matrix: Nitric Acid (tr. Tartaric Acid) (HNO3 / Tartaric Acid), Dilution: 1:1000, Analyte: Sb 500 ug/mL.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-520 Supplier Inorganic Ventures Supplier No. CLPPSPK2125ML
Matrix: Nitric Acid (tr. Tartaric Acid) (HNO3 / Tartaric Acid), Dilution: 1:1000, Analyte: Sb 500 ug/mL.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search