Learn More
Inorganic Ventures ICP CALIBRATION STD 35 125mL
SDP
Catalog No. 50272313
Encompass_Preferred
Matrix: Nitric Acid (HNO3), Instrument(s): Perkin Elmer, Analyte: Ca, K, Fe, Mg, and Na 1000 ug/mL each.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-272-313 Supplier Inorganic Ventures Supplier No. IVSTOCK35125ML
Add to Cart
Add to Cart
Matrix: Nitric Acid (HNO3), Instrument(s): Perkin Elmer, Analyte: Ca, K, Fe, Mg, and Na 1000 ug/mL each.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search