Learn More
Inorganic Ventures ICP CALIBRATION STD 36 500mL
SDP
Catalog No. 50272314
Encompass_Preferred
Inorganic Ventures' ICP Calibration Standard 36 is a 500mL certified reference material set in a hydrochloric acid matrix for stability. This standard is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. See Certificate of Analysis for expiration.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-272-314 Supplier Inorganic Ventures Supplier No. IVSTOCK36500ML
Inorganic Ventures' ICP Calibration Standard 36 is a 500mL certified reference material set in a hydrochloric acid matrix for stability. This standard is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. See Certificate of Analysis for expiration.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search