Learn More
Alkali Scientific ICP MS Autosampler Tube, Polypropylene (PP) – 16mL (17 × 130mm), 100/Pk
SDP
Catalog No. 5000661952
Encompass_Preferred
High purity polypropylene (PP) test tubes designed for ICP MS (Inductively Coupled Plasma Mass Spectrometry) and trace metal analysis. These tubes help minimize contamination and ensure reproducible results in ultra trace elemental testing. They are ideal for laboratories that require high sample integrity in environmental, clinical, and industrial applications. ICP MS analysis is commonly used for environmental monitoring of trace metals in water, soil, and air samples; food and drug testing for elemental impurities; life sciences and clinical research for sample digestion and elemental quantification; and mining or geochemistry for ore and mineral composition analysis. Manufactured from ultra clean, trace metal free polypropylene, each tube has a 16 mL capacity and dimensions of 17 × 130 mm. Supplied in packs of 100 tubes and compatible with most ICP MS autosamplers and sample handling systems.
Catalog No. 50-006-61952 Supplier Alkali Scientific Supplier No. ICP00
High purity polypropylene (PP) test tubes designed for ICP MS (Inductively Coupled Plasma Mass Spectrometry) and trace metal analysis. These tubes help minimize contamination and ensure reproducible results in ultra trace elemental testing. They are ideal for laboratories that require high sample integrity in environmental, clinical, and industrial applications. ICP MS analysis is commonly used for environmental monitoring of trace metals in water, soil, and air samples; food and drug testing for elemental impurities; life sciences and clinical research for sample digestion and elemental quantification; and mining or geochemistry for ore and mineral composition analysis. Manufactured from ultra clean, trace metal free polypropylene, each tube has a 16 mL capacity and dimensions of 17 × 130 mm. Supplied in packs of 100 tubes and compatible with most ICP MS autosamplers and sample handling systems.