Learn More
Perkin Elmer US LLC 9-Element Solution, 2% HNO3, 10 µg/mL, 125 mL
Supplier:  Perkin Elmer US LLC N9300231
Encompass_Preferred
Details Concentration Element 10 µg/mL Be, Bi, Ce, Co, In, Mg, Ni, Pb, U
Catalog No. 50-904-5749