Learn More
AccuStandard, Inc ICP MS CAL STD 2 METH 200.8
SDP
Catalog No. NC9180519
Encompass
ICP-MS Calibration Standard 2 for Method 200.8. 100 mL. 1000 ?g/mL in 2-5% Nitric Acid.
Catalog No. NC9180519 Supplier AccuStandard, Inc Supplier No. ICPMS200.8CAL21
ICP-MS Calibration Standard 2 for Method 200.8. 100 mL. 1000 ?g/mL in 2-5% Nitric Acid.