Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ICP-MS Calibration Standard 1R, Method 200.8 (94), Ricca Chemical
SDP

Catalog No. RMSCAL1R100 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RMSCAL1R-100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RMSCAL1R-100 Supplier Ricca Chemical Company Supplier No. RMSCAL1R100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material ICP-MS Calibration Standard 1R
Packaging Natural LDPE Bottle
CAS 7732-18-5, 7697-37-2, 7789-02-8, 7789-02-8, 7782-49-2, 7664-39-3
CAS Min % 93.0, 2.94, 0.01, 0.01, 0.0047, 0.0029
CAS Max % 97.0, 3.06, 0.01, 0.01, 0.0049, 0.0031
Color Colorless
pH Acidic
Boiling Point Approximately 100°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type ICP Multi-element Standard Solution
Quantity 100 mL
Grade Laboratory
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.