Learn More
Agilent Technologies ICP-MS Stock Tuning Solution, Multi-Element Mixture, 100 mL
Catalog No. NC3162589
Encompass
This stock tuning solution is a multi-element calibration standard designed for Inductively Coupled Plasma Mass Spectrometry (ICP-MS) applications. It contains lithium, yttrium, cerium, thallium, and cobalt, each at a concentration of 10 μg/mL, formulated in a 2% nitric acid matrix. This ready-to-use mixture simplifies the tuning process for analytical instruments, ensuring precise and reproducible performance for routine laboratory analysis.
  • Ready-to-use multi-element tuning standard reduces preparation time compared with single-element solutions
  • Formulated in 2% HNO3 for direct compatibility with ICP-MS sample introduction systems
  • ISO 17034 quality designation supports consistent, certified composition for reproducible tuning and calibration
  • 100 mL volume convenient for routine laboratory use
Catalog No. NC3162589 Supplier Agilent Technologies Supplier No. 51886564
This stock tuning solution is a multi-element calibration standard designed for Inductively Coupled Plasma Mass Spectrometry (ICP-MS) applications. It contains lithium, yttrium, cerium, thallium, and cobalt, each at a concentration of 10 μg/mL, formulated in a 2% nitric acid matrix. This ready-to-use mixture simplifies the tuning process for analytical instruments, ensuring precise and reproducible performance for routine laboratory analysis.
  • Ready-to-use multi-element tuning standard reduces preparation time compared with single-element solutions
  • Formulated in 2% HNO3 for direct compatibility with ICP-MS sample introduction systems
  • ISO 17034 quality designation supports consistent, certified composition for reproducible tuning and calibration
  • 100 mL volume convenient for routine laboratory use